Learn More
Abeba™ Dark Blue Velours ESD Anatom Safety Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand: Abeba™ 32147/46
Additional Details : Weight : 1.77800kg
Description
- CE EN ISO 20345:2011 S1 SRC
- Velours with breathable textile inlays
- Dark blue
- Steel toe cap
- Shock absorbant PU midsole
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3580)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
- Suitable for heel and foot lift
- Scratch-resistant coating in heel area
Specifications
CE EN ISO 20345:2011 S1 SRC | |
Velour | |
Slip-resistant | |
32147 |
Blue | |
46 | |
Polyurethane | |
Steel |