missing translation for 'onlineSavingsMsg'
Learn More

SCHOTT AG Lighting and Imaging™ Darkfield Optic Ring for VisiLED Slim LED Ring Light

Darkfield Optic Ring for VisiLED Slim LED Ring Light provides working distance from 5 to 25 mm.

Brand:  SCHOTT AG Lighting and Imaging™ 400810

Product Code. 15773042

  • £204.00 / 1 unit
Estimated Shipment: 10-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Focus optics ring for darkfield.
  • Working distance 5 to 25 mm.
  • For use with VisiLED Slim Ringlight (400 150).
Specifications

Specifications

Darkfield Optic Ring
Free working distance: 5 to 20 mm
Stereo Microscopy Illumination
VisiLED Slim LED Ringlight (400 150)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SCHOTT AG Lighting and Imaging™ Darkfield Optic Ring for VisiLED Slim LED Ring Light > Darkfield Optic Ring; For Use With: VisiLED Slim LED Ring Light; Focus Distance: 5 to 25 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.