missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Darunavir, Thermo Scientific™
Click to view available options
Quantity:
100 mg
Specifications
CAS | 206361-99-1 |
Packaging | Glass Bottle |
Quantity | 100 mg |
Color | White |
Certificate of Analysis | Available |
Molecular Formula | C27H37N3O7S |
MDL Number | MFCD09260006 |
InChI Key | CJBJHOAVZSMMDJ-HEXNFIEUSA-N |
SMILES | CC(C)CN(C[C@@H](O)[C@H](CC1=CC=CC=C1)NC(=O)O[C@H]1CO[C@H]2OCC[C@@H]12)S(=O)(=O)C1=CC=C(N)C=C1 |
IUPAC Name | (3R,3aS,6aR)-hexahydrofuro[2,3-b]furan-3-yl N-[(2S,3R)-3-hydroxy-4-[N-(2-methylpropyl)4-aminobenzenesulfonamido]-1-phenylbutan-2-yl]carbamate |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'