missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Dasatinib

Product Code. 17120172
Encompass_Preferred
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
This item is not returnable. View return policy

Product Code. 17120172

Brand: STEMCELL Technologies 73084

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tyrosine kinase inhibitor; Inhibits ABL, SRC, LCK, and YES

  • Dasatinib is a potent, ATP-competitive tyrosine kinase inhibitor
  • It is specific for SRC/ABL kinases, for example, ABL, SRC, LCK, and YES with IC50 values of < 1.0, 0.5, 0.4 and 0.5 nM, respectively, and also demonstrates activity against KIT with an IC50 = 5.0 nM (Lombardo et al.; Davis et al.) Dasatinib is a second-generation inhibitor of the oncogenic tyrosine kinase BCR-ABL with 325-fold more potency than imatinib, and is also able to inhibit imatinib-resistant BCR-ABL mutants (Tokarski et al.) It also inhibits a large number of other kinases (76 of 148 kinases tested) when screened at 10 μM (Carter et al.)

Specifications

CAS 302962-49-8
Synonym BMS 354825; Sprycel
Quantity 100 mg
Molecular Formula C22H26ClN7O2S
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.