missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech
This item is not returnable.
View return policy
Description
The Dopamine Transporter (DAT) is responsible for the reaccumulation of dopamine after it has been released. DAT antibodies and antibodies for other markers of catecholamine biosynthesis are widely used as markers for dopaminergic and noradrenergic neurons in a variety of applications including depression, schizophrenia, Parkinson′s disease and drug abuse.Specifications
DAT | |
Polyclonal | |
Unconjugated | |
SLC6A3 | |
DA transporter, DAT, DAT1, SLC6A3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13162, 24898, 6531 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P23977, Q01959, Q61327 | |
SLC6A3 | |
DAT Fusion Protein Ag18297 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |