Learn More
Fisherbrand™ Traceable™ Data Acquisition Software
Shop All Cole-Parmer Products
Click to view available options
Includes:
Cailbration, Software CD and 6 ft. Cables (USB, Serial, and Instrument Connections)
Unit Size:
Each
Description
- Display and store measurement readings on a PC from up to 8 devices at a time
- Data capture runs in the background, enabling users to work on other programs simultaneously
- Set audible and visual notification alarms, or immediately send an email if an over/under condition occurs
- Software can be used to connect a meter directly to PC via cable
- Check software specification tables for meter compatibility
Specifications
Specifications
| Type | Data Acquisition Software |
| Display Type | No Display |
| Description | Data Acquisition Software with USB Cable and Calibration |
| Includes | Cailbration, Software CD and 6 ft. Cables (USB, Serial, and Instrument Connections) |
| For Use With (Equipment) | 90080-09, 03313-65, 03313-66, 37804-06, 37804-07 (Traceable™ 4000, 4080, 4085, 6412, 6413) |