Learn More
Sartorius Stedim Biotech™ Data Communication Cable for Laboratory Balances
Sartorius Stedim Biotech™ Data Communication Cable for Laboratory Balances is suitable for a variety of laboratory balances.
Brand: Sartorius Stedim Biotech™ YCC03-D25
Code : NEW
Additional Details : Weight : 0.02000kg
Description
- Suitable for the following laboratory balances:
- Secura™
- Quntix™
- Practum™
- Mini USB to RS232. 25-pin
Specifications
Secura, Quintix, Practum Laboratory Balance | |
USB/RS232 25-pin connectors | |
USB/RS232 |