missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Data Output Accessories for Environmental Chambers and Incubators
Integrate Thermo Scientific™ Data Output Accessories for your environmental chamber or incubator to increase functionality and compatibility.
Brand: Thermo Scientific™ 201144
This item is not returnable.
View return policy
Description
Integrate Thermo Scientific™ Data Output Accessories for your environmental chamber or incubator to increase functionality and compatibility.
Specifications
Single pen, 6 in. 7 Day Chart Recorders; 0° to 60°C (32° to 140°F) | |
LED | |
Accessory, Chart Recorders and Paper |
Environmental Chamber (3948 and 3907) | |
0°C to 60°C | |
Factory Installed |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction