Learn More
Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers
Integrate Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers with a variety of ultra-low temperature freezers to increase functionality and compatibility.
£377.75 - £500.80
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11973937
|
Thermo Scientific™
195895 |
RS-232; 0-1V |
£377.75
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
11983937
|
Thermo Scientific™
195896 |
RS-232; 0-5V |
£386.36
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
11796649
|
Thermo Scientific™
195897 |
RS-485; 4-20mA |
£475.85
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
11913947
|
Thermo Scientific™
195898 |
RS-485; 0-1V |
£498.22
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
11339301
|
Thermo Scientific™
195899 |
RS-485; 0-5V |
£500.80
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
Description
Integrate Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers with a variety of ultra-low temperature freezers to increase functionality and compatibility.
RS-232:- RS-232 and 4-40mA analog outputs are standard on Thermo Scientific™ Forma™ 8600 Series freezers. These additional options are designed to meet specific needs.
- RS-232 allows connection to a computer, recorder, or independent monitoring device RS-485 allows direct connection to the Model 1535 Monitor/Alarm System
Note:
RS-232 is not compatible with the Forma Model 1535 Monitor/Alarm System.
Specifications
Alarm, Recorder |