missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Davercin, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
Description
Davercin (Erythromycin Cyclocarbonate), derivative of Erythromycin, which is active against Gram-positive and some Gram-negative microorganisms.
Specifications
Specifications
| Chemical Name or Material | Davercin |
| CAS | 55224-05-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C38H65NO14 |
| Quantity | 5 mg |
| Synonym | Erythromycin Cyclocarbonate |
| Solubility Information | DMSO : ≥ 50 mg/mL (65.80 mM) |
| SMILES | C[C@]([C@H](OC([C@@H]([C@H]([C@H](C)[C@H]1O[C@@](O[C@H](C)C[C@@H]2N(C)C)([H])[C@@H]2O)O[C@@](O[C@@H](C)[C@@H]3O)([H])C[C@@]3(C)OC)C)=O)CC)(O4)[C@]([C@H](C([C@@H](C[C@]1(O)C)C)=O)C)([H])OC4=O |
| Molecular Weight (g/mol) | 759.92 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction