Learn More
SCHOTT AG Lighting and Imaging™ Daylight Filter for Halogen Light Sources
The Daylight Filter for Halogen Light Sources Increases the color temperature from a warm halogen light to a cold daylight. For use with focusing lens with 5 mm diameter light guide and KL1500HAL Cold Light Source.
Brand:  SCHOTT AG Lighting and Imaging™ 158211
Additional Details : Weight : 5.00000kg
Product Code. 15773022
  • £37.86 / 1 unit
Estimated Shipment: 10-09-2024
to see stock.
Add to basket
Description
Description
For use with:
  • Focusing lens with 5 mm diameter light guide
  • KL1500HAL Cold Light Source
Specifications
Specifications
Filter
Stereo Microscopy Illumination
Focusing Lens 158210, KL1500HAL
Daylight