missing translation for 'onlineSavingsMsg'
Learn More

DCA, Tocris Bioscience™

Product Code. 18732694
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Mitochondrial pyruvate dehydrogenase kinase (PDK) inhibitor

Inhibitor of mitochondrial pyruvate dehydrogenase kinase (PDK). Shifts pyruvate metabolism from glycolysis and lactate production to glucose oxidation in the mitochondria. Induces mitochondrial-dependent apoptosis and reverses the inhibition/downregulation of KV1.5 channels in cancer but not normal cells. Decreases tumor growth in vitro and in vivo.

Specifications

CAS 2156-56-1
Quantity 100 mg
Molecular Formula C2HCl2NaO2
MDL Number MFCD00070489
Synonym sodium dichloroacetate, sodium 2,2-dichloroacetate, dichloroacetic acid sodium salt, acetic acid, dichloro-, sodium salt, ceresine, dichloroacetate sodium, dichloroctan sodny czech, sodium dichloroacetate usan, ccris 7697, dichloroacetic acid, sodium salt
InChI Key LUPNKHXLFSSUGS-UHFFFAOYSA-M
SMILES [Na+].[O-]C(=O)C(Cl)Cl
IUPAC Name sodium 2,2-dichloroacetate
Molecular Weight (g/mol) 150.92
PubChem CID 517326
Formula Weight 150.92
Percent Purity >98%
Chemical Name or Material DCA
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.