Learn More
DCTPP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16684-1-AP-20UL
 View more versions of this product
Product Code. 16836394
  • £135.00 / 20µL
Explore more special offers
Description
Description
The protein encoded by this gene is dCTP pyrophosphatase, which converts dCTP to dCMP and inorganic pyrophosphate. The encoded protein also displays weak activity against dTTP and dATP, but none against dGTP. This protein may be responsible for eliminating excess dCTP after DNA synthesis and may prevent overmethylation of CpG islands. Three transcript variants, one protein-coding and the other two non-protein coding, have been found for this gene.
Specifications
Specifications
DCTPP1
Polyclonal
Unconjugated
DCTPP1
CDA03, dCTP pyrophosphatase 1, dCTPase 1, DCTPP1, Deoxycytidine triphosphatase 1, RS21C6, XTP3TPA
Rabbit
Antigen Affinity Chromatography
RUO
192252, 66422, 79077
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunofluorescence
0.38 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q91VC0, Q9H773, Q9QY93
Dctpp1
DCTPP1 Fusion Protein Ag10015
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote