missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DDR2/CEN1p FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
£1235.00 - £2030.00
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | discoidin domain receptor tyrosine kinase 2 |
Gene Alias | MIG20a,NTRKR3,TKT,TYRO10 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16053325
|
Abnova™
FG0203.100uL |
100 μL |
£1235.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16063325
|
Abnova™
FG0203.200uL |
200 μL |
£2030.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
MIG20a,NTRKR3,TKT,TYRO10 | |
4921 | |
Liquid |
Tube | |
discoidin domain receptor tyrosine kinase 2 | |
DDR2 | |
DNA/RNA |