missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Dead Cell Apoptosis Kits with Annexin V for Flow Cytometry

Assess cell viability with Dead Cell Apoptosis kits with Annexin V and conjugated fluorescent dyes such as Alexa Fluor 488, FITC, PI, PE, APC, and SYTOX Green.

Brand:  Molecular Probes™ V13242

267.33 GBP valid until 2025-06-30
Use promo code "24074" to get your promotional price.


 View more versions of this product

Product Code. 10267392

  • £399.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Dead Cell Apoptosis Kit with Annexin V FITC and PI, for flow cytometry
Contains 1 vial of annexin V, FITC conjugate (250 μL ), 1 vial of propidium iodide (PI, 100 μL), and 1 bottle of annexin binding buffer (5X solution, 15 mL).
Store in refrigerator (2°C to 8°C) and protect from light.
494, 535/518, 617
Wet Ice
FITC, Propidium Iodide
1 Kit
Apoptosis Detection Kit
488
50
Flow Cytometer
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2975948 Certificate TypeCertificate of Analysis Date16/10/2024 Catalog NumberV13242
Lot Number2720247 Certificate TypeCertificate of Analysis Date02/12/2023 Catalog NumberV13242
Lot Number2541545 Certificate TypeCertificate of Analysis Date02/02/2023 Catalog NumberV13242
Lot Number2445381 Certificate TypeCertificate of Analysis Date18/03/2022 Catalog NumberV13242
Lot Number2354351 Certificate TypeCertificate of Analysis Date23/07/2021 Catalog NumberV13242
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular Probes™ Dead Cell Apoptosis Kits with Annexin V for Flow Cytometry > 1 kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.