missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decacarbonyldirhenium, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
Decacarbonyldirhenium is a key starting material used for the synthesis of other rhenium carbonyl complexes. It plays an important role in metathesis, reforming and hydrogenation reactions. It is used as a catalyst to promote silation of alcohols and in the preparion of silyl ethers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 170°C (decomposition) |
Color | White |
Density | 2.87 g/mL |
Linear Formula | Re2(CO)10 |
Quantity | 1 g |
UN Number | UN3466 |
IUPAC Name | carbon monoxide;rhenium |
Formula Weight | 652.52 |
Percent Purity | 96% |
Physical Form | Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'