missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decahydronaphthalene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
62.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
5 mL
500 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -31.0°C |
Density | 0.8900g/mL |
Boiling Point | 187.0°C |
Flash Point | 58°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | C10H18 |
Refractive Index | 1.4740 to 1.4760 |
Quantity | 2.5 L |
Show More |
Safety and Handling
Product Identifier
- Decahydronaphthalene, mixture of cis and trans
- Danger
- Flammable liquid Category 3
- Aspiration toxicity Category 1
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 C
- LONG-TERM AQUATIC HAZARD Chronic 2
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'