Learn More
Decahydronaphthalene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
£22.21 - £519.59
Chemical Identifiers
CAS | 91-17-8 |
---|---|
Molecular Formula | C10H18 |
Molecular Weight (g/mol) | 138.25 |
MDL Number | MFCD00004130 |
InChI Key | NNBZCPXTIHJBJL-UHFFFAOYSA-N |
Synonym | decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane |
PubChem CID | 7044 |
ChEBI | CHEBI:38853 |
IUPAC Name | 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene |
SMILES | C1CCC2CCCCC2C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10487290
|
Thermo Scientific Acros
111840050 |
5 mL | Glass bottle |
£22.21
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10315890
|
Thermo Scientific
111845000 |
500 mL | Glass bottle |
£26.03
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10540191
|
Thermo Scientific Acros
111840010 |
1 L | Glass bottle |
£41.61
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10606492
|
Thermo Scientific Acros
111840025 |
2.5 L | Glass bottle |
£76.98
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10792764
|
Thermo Scientific Acros
111840250 |
25 L | Metal Drum |
£519.59
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
91-17-8 | |
138.25 | |
NNBZCPXTIHJBJL-UHFFFAOYSA-N | |
7044 | |
1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene |
C10H18 | |
MFCD00004130 | |
decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane | |
CHEBI:38853 | |
C1CCC2CCCCC2C1 |
Specifications
91-17-8 | |
0.8900g/mL | |
58°C | |
97.5% min. (GC) | |
C10H18 | |
MFCD00004130 | |
15, 2850 | |
decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane | |
NNBZCPXTIHJBJL-UHFFFAOYSA-N | |
1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene | |
7044 | |
3 mPa.s (20°C) | |
98% | |
Decahydronaphthalene, Mixture of cis and trans |
-31.0°C | |
187.0°C | |
Authentic | |
C10H18 | |
1.4740 to 1.4760 | |
05, III, 245 | |
0.89 | |
Solubility in water: slightly soluble (6mg/l at 20°C). Other solubilities: very soluble in acetone,benzene,ether and,ethanol | |
C1CCC2CCCCC2C1 | |
138.25 | |
CHEBI:38853 | |
138.25 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-046-9
RUO – Research Use Only