missing translation for 'onlineSavingsMsg'
Learn More

Decahydronaphthalene, Mixture of cis + trans, 98%, Reagent Grade, Decalin™, Honeywell™

Product Code. 15657810
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Aluminum bottle
Glass bottle
Stainless steel can
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-17-8
Molecular Formula C10H18
Molecular Weight (g/mol) 138.254
MDL Number MFCD00004130
InChI Key NNBZCPXTIHJBJL-UHFFFAOYSA-N
Synonym decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane
PubChem CID 7044
ChEBI CHEBI:38853
IUPAC Name 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene
SMILES C1CCC2CCCCC2C1

Specifications

Boiling Range 189° - 191°C
Melting Point -125°C
Density 0.896g/mL at 25°C
Explosion Limit 0.7 to 4.9%, 100°F
Flash Point 57°C (134.6°F)
Ignition Point 482°F
Packaging Glass bottle
Quantity 500 mL
UN Number UN1147
Beilstein 878165
Vapor Density 4.76 (vs air)
Formula Weight 138.25g/mol
Vapor Pressure 42mmHg (92°C), 741mmHg (188°C)
Percent Purity 0.98
Chemical Name or Material Decahydronaphthalene, mixture of cis plus trans
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'