missing translation for 'onlineSavingsMsg'
Learn More

Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals

£25.43 - £86.93

Chemical Identifiers

CAS 541-02-6
Molecular Formula C10H30O5Si5
Molecular Weight (g/mol) 370.77
InChI Key XMSXQFUHVRWGNA-UHFFFAOYSA-N
Synonym decamethylcyclopentasiloxane, cyclopentasiloxane, decamethyl, cyclomethicone 5, dimethylsiloxane pentamer, dow corning 345 fluid, nuc silicone vs 7158, dow corning 345, silicon sf 1202, cyclic dimethylsiloxane pentamer, dekamethylcyklopentasiloxan
PubChem CID 10913
IUPAC Name 2,2,4,4,6,6,8,8,10,10-decamethyl-1,3,5,7,9,2,4,6,8,10-pentaoxapentasilecane
SMILES C[Si]1(O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15289936
View Documents Promotion Details
Thermo Scientific Acros
454580500
50 g
£25.43
50g
Please to purchase this item. Need a web account? Register with us today!
15299936
View Documents
Thermo Scientific Acros
454582500
250 g
£86.93
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

541-02-6
370.77
decamethylcyclopentasiloxane, cyclopentasiloxane, decamethyl, cyclomethicone 5, dimethylsiloxane pentamer, dow corning 345 fluid, nuc silicone vs 7158, dow corning 345, silicon sf 1202, cyclic dimethylsiloxane pentamer, dekamethylcyklopentasiloxan
2,2,4,4,6,6,8,8,10,10-decamethyl-1,3,5,7,9,2,4,6,8,10-pentaoxapentasilecane
C10H30O5Si5
XMSXQFUHVRWGNA-UHFFFAOYSA-N
10913
C[Si]1(O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)C

Specifications

541-02-6
101.0°C (20.0 mmHg)
96% min. (GC)
15,2852
decamethylcyclopentasiloxane, cyclopentasiloxane, decamethyl, cyclomethicone 5, dimethylsiloxane pentamer, dow corning 345 fluid, nuc silicone vs 7158, dow corning 345, silicon sf 1202, cyclic dimethylsiloxane pentamer, dekamethylcyklopentasiloxan
XMSXQFUHVRWGNA-UHFFFAOYSA-N
2,2,4,4,6,6,8,8,10,10-decamethyl-1,3,5,7,9,2,4,6,8,10-pentaoxapentasilecane
10913
370.77
Decamethylcyclopentasiloxane
0.9590g/mL
76°C
C10H30O5Si5
0.959
Solubility in water: insoluble
C[Si]1(O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)C
370.77
4 mPa.s (20°C)
97%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 208-764-9

Product Identifier
  • Decamethylcyclopentasiloxane

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only