missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decane, 99+%, pure
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| Density | (15°C) 0.733 to 0.737kg/L |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99+% |
| Packaging | Glass bottle |
| Linear Formula | CH3(CH2)8CH3 |
| Refractive Index | 1.4103 to 1.4123 (20°C, 589nm) |
| Quantity | 100 mL |
| Beilstein | 01, 168 |
| Formula Weight | 142.28 |
| Show More |
Safety and Handling
Product Identifier
- Decane
- Danger
- Aspiration toxicity Category 1
- Flammable liquid Category 3
- H304-May be fatal if swallowed and enters airways.
- H226-Flammable liquid and vapour.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto