missing translation for 'onlineSavingsMsg'
Learn More

Decanoic acid, 99%, Thermo Scientific Chemicals

£25.93 - £359.79

Chemical Identifiers

CAS 334-48-5
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
MDL Number MFCD00004441
InChI Key GHVNFZFCNZKVNT-UHFFFAOYSA-N
Synonym capric acid, n-decanoic acid, n-capric acid, decylic acid, caprinic acid, decoic acid, n-decylic acid, n-decoic acid, 1-nonanecarboxylic acid, caprynic acid
PubChem CID 2969
ChEBI CHEBI:30813
IUPAC Name decanoic acid
SMILES CCCCCCCCCC(=O)O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10114090
View Documents Promotion Details
Thermo Scientific Acros
167271000
100 g Glass bottle
£25.93
100g
Please to purchase this item. Need a web account? Register with us today!
10722211
View Documents Promotion Details
Thermo Scientific Acros
167275000
500 g Glass bottle
£93.87
500g
Please to purchase this item. Need a web account? Register with us today!
10014823
View Documents Promotion Details
Thermo Scientific Acros
167270010
1 kg Glass bottle
£167.84
1kg
Please to purchase this item. Need a web account? Register with us today!
10465125
View Documents Promotion Details
Thermo Scientific Acros
167270025
2.5 kg Plastic Drum
£359.79
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

334-48-5
172.268
GHVNFZFCNZKVNT-UHFFFAOYSA-N
2969
decanoic acid
C10H20O2
MFCD00004441
capric acid, n-decanoic acid, n-capric acid, decylic acid, caprinic acid, decoic acid, n-decylic acid, n-decoic acid, 1-nonanecarboxylic acid, caprynic acid
CHEBI:30813
CCCCCCCCCC(=O)O

Specifications

334-48-5
268.0°C to 270.0°C
99%
CH3(CH2)8CO2H
02, II, 309
capric acid, n-decanoic acid, n-capric acid, decylic acid, caprinic acid, decoic acid, n-decylic acid, n-decoic acid, 1-nonanecarboxylic acid, caprynic acid
GHVNFZFCNZKVNT-UHFFFAOYSA-N
decanoic acid
2969
172.27
0.8900g/mL
Authentic
0.89
30.0°C to 32.0°C
White
C10H20O2
MFCD00004441
15, 1760
Solubility in water: 0.15g/L (20°C). Other solubilities: soluble in dilute hno3, soluble in ethanol, ether, chloroform, benzene and, carbon disulfide
CCCCCCCCCC(=O)O
172.268
CHEBI:30813
99%
150°C
Crystalline Solid
Decanoic acid, 99+%
Videos
Safety and Handling

Safety and Handling

GHS P Statement
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

missing translation for 'einecsNumber' : 206-376-4

missing translation for 'rtecsNumber' : HD9100000

missing translation for 'tsca' : TSCA

Product Identifier
  • Decanoic acid

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P362-Take off contaminated clothing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only