Learn More
Decanoyl chloride, 98+%, Thermo Scientific Chemicals
£32.00 - £124.00
Chemical Identifiers
CAS | 112-13-0 |
---|---|
Molecular Formula | C10H19ClO |
Molecular Weight (g/mol) | 190.71 |
MDL Number | MFCD00000771 |
InChI Key | IPIVAXLHTVNRBS-UHFFFAOYSA-N |
Synonym | capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n |
PubChem CID | 66982 |
IUPAC Name | decanoyl chloride |
SMILES | CCCCCCCCCC(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10081880
|
Thermo Scientific Acros
161191000 |
100 g | Glass bottle |
£32.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10781431
|
Thermo Scientific Acros
161195000 |
500 g | Glass bottle |
£124.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-13-0 | |
190.71 | |
IPIVAXLHTVNRBS-UHFFFAOYSA-N | |
66982 | |
CCCCCCCCCC(=O)Cl |
C10H19ClO | |
MFCD00000771 | |
capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n | |
decanoyl chloride |
Specifications
112-13-0 | |
Brown to White | |
232.0°C | |
Authentic | |
C10H19ClO | |
1.4400 to 1.4420 | |
02, 356 | |
capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n | |
IPIVAXLHTVNRBS-UHFFFAOYSA-N | |
decanoyl chloride | |
66982 | |
98+% | |
Decanoyl chloride, 98% |
-34.5°C | |
0.9190g/mL | |
110°C | |
98% min. (GC) | |
CH3(CH2)8COCl | |
MFCD00000771 | |
0.919 | |
Solubility in water: 100mg/l (20°C). Other solubilities: hydrolyses in water,450g/L methanol- 500g/L ethanol,500g/L 2-propanol- 500g/L dmf,500g/L acetone- 500g/L dichloromethane | |
CCCCCCCCCC(=O)Cl | |
190.71 | |
190.71 | |
Powder |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-938-0
missing translation for 'tsca' : TSCA
- Decanoyl chloride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only