Learn More
Decanoyl chloride, 98%, Thermo Scientific Chemicals
£28.90 - £96.10
Chemical Identifiers
CAS | 112-13-0 |
---|---|
Molecular Formula | C10H19ClO |
Molecular Weight (g/mol) | 190.711 |
MDL Number | MFCD00000771 |
InChI Key | IPIVAXLHTVNRBS-UHFFFAOYSA-N |
Synonym | capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n |
PubChem CID | 66982 |
IUPAC Name | decanoyl chloride |
SMILES | CCCCCCCCCC(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11484057
|
Thermo Scientific Alfa Aesar
A19486.22 |
100 g |
£28.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494057
|
Thermo Scientific Alfa Aesar
A19486.36 |
500 g |
£96.10
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Decanoyl chloride is used in the synthesis of oligomers composed of alternating 2,6-diaminopyridine and 2,6-pyridinedicarbonyl units. It enhances the activity of proleather from Bacillus sp. during the synthesis of poly(lactic acid) in organic solvents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDecanoyl chloride is used in the synthesis of oligomers composed of alternating 2,6-diaminopyridine and 2,6-pyridinedicarbonyl units. It enhances the activity of proleather from Bacillus sp. during the synthesis of poly(lactic acid) in organic solvents.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Chemical Identifiers
112-13-0 | |
190.711 | |
IPIVAXLHTVNRBS-UHFFFAOYSA-N | |
66982 | |
CCCCCCCCCC(=O)Cl |
C10H19ClO | |
MFCD00000771 | |
capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n | |
decanoyl chloride |
Specifications
112-13-0 | |
0.925 | |
106°C (223°F) | |
C10H19ClO | |
MFCD00000771 | |
507055 | |
capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n | |
IPIVAXLHTVNRBS-UHFFFAOYSA-N | |
decanoyl chloride | |
66982 | |
98% |
-35°C | |
232°C | |
Irritating | |
1.441 | |
UN3265 | |
Moisture sensitive | |
Reacts with water. | |
CCCCCCCCCC(=O)Cl | |
190.711 | |
190.71 | |
Decanoyl chloride |
Safety and Handling
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P234-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 203-938-0
missing translation for 'rtecsNumber' : HE0875000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only