missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decitabine, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Decitabine (NSC 127716) is an orally active deoxycytidine analogue antimetabolite and a DNA methyltransferase inhibitor. Decitabine incorporates into DNA in place of cytosine can covalently trap DNA methyltransferase to DNA causing irreversible inhibition of the enzyme. Decitabine induces cell G2/M arrest and cell apoptosis. Decitabine has potent anticancer activity.
Specifications
Specifications
| Chemical Name or Material | Decitabine |
| CAS | 2353-33-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C8H12N4O4 |
| Quantity | 50 mg |
| Synonym | 5-Aza-2'-deoxycytidine 5-AZA-CdR NSC 127716 |
| Solubility Information | DMSO : ≥ 50 mg/mL (219.10 mM) ∣H2O : 20 mg/mL (87.64 mM; Need ultrasonic) |
| SMILES | O=C1N=C(N)N=CN1[C@H](O[C@@H]2CO)C[C@@H]2O |
| Molecular Weight (g/mol) | 228.21 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction