missing translation for 'onlineSavingsMsg'
Learn More

Decon™ Decomatic

Biodegradable, phosphate, enzyme and chlorine free, suitable for automatic washing machines

Brand:  Decon™ DEC10


 View more versions of this product

Product Code. 10538610

  • £251.00 / 10 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Biodegradable, phosphate, enzyme and chlorine free, suitable for automatic washing machines

Biodegradable, phosphate, enzyme and chlorine free, suitable for automatic washing machines. By preventing the deposition of lime and salts on both contents and machine parts, it minimises corrosion and wear of seals, pumps, racks and stainless steel surfaces, thereby providing more efficient performance and longer life to the machine. Not suitable for use on non-ferrous metals, notably aluminum and zinc, or on polycarbonate. Ideal for use on glassware and equipment in medical, pharmaceutical, research, educational, scientific and industrial laboratories.
Specifications

Specifications

Liquid
Amber
2.28cps at 20°C
>13
Phosphate, Enzyme-Free, Chlorine-Free
10 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • DECON DECOMATIC

Signal Word
  • Warning

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Supplemental information
  • MIXTURE LIST-Contains : NaOH

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Decon™ Decomatic > Quantity: 10,000mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.