Learn More
Thermo Scientific™ Decontamination Kit
For Use With: For Water Jacket CO2 Incubators
Brand:  Thermo Scientific™ LT190651
Product Code. 10616240
  • £214.00 / Each
Estimated Shipment: 11-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For Use With: For Water Jacket CO2 Incubators
HEPA Filters
  • 99.97% filtering at 0.3μm
  • Compatible with all Thermo Scientific™ Forma™ Series II 3110, Thermo Scientific™ Steri-Cycle™, and Forma™ 310 Series (with HEPA filtration system option) incubators
  • Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
Decontamination Kits
  • Consists of filters for difficult-to-clean chambers and gaskets (except door) and tubing
  • Reduces downtime and labor spent on decontamination
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Internal chamber replacement kit with HEPA, gaskets, and other parts
For Water Jacket CO2 Incubators
LED
Decontamination Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Decontamination Kit > Decon Kit; For 3110 Series Incubators