missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decyldimethylchlorosilane, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Specifications
Specifications
Color | Colorless |
Density | 0.8660g/mL |
Boiling Point | 98°C (3.0 hPa) |
Flash Point | 137°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.4400 to 1.4420 (20°C, 589nm) |
Quantity | 5 mL |
Specific Gravity | 0.866 |
Show More |
Safety and Handling
Product Identifier
- Decyldimethylchlorosilane, 97%
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH029-Contact with water liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'