missing translation for 'onlineSavingsMsg'
Learn More

Decyltrimethylammonium bromide, 99%

Product Code. 10106800 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10106800

Brand: Thermo Scientific Acros 406230250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2082-84-0 | C13H30BrN | 280.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2082-84-0
Molecular Formula C13H30BrN
Molecular Weight (g/mol) 280.29
MDL Number MFCD00041973
InChI Key PLMFYJJFUUUCRZ-UHFFFAOYSA-M
Synonym decyltrimethylammonium bromide, n,n,n-trimethyldecan-1-aminium bromide, n-decyltrimethylammonium bromide, trimethyldecylammonium bromide, n,n,n-trimethyldecylammonium bromide, n,n,n-trimethyl-1-decanaminium bromide, 1-decanaminium, n,n,n-trimethyl-, bromide, decyl trimethyl azanium bromide, fsm 20, decyl trimethyl ammonium bromide
PubChem CID 16388
ChEBI CHEBI:295756
IUPAC Name decyl(trimethyl)azanium;bromide
SMILES [Br-].CCCCCCCCCC[N+](C)(C)C

Specifications

Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula CH3(CH2)9N(CH3)3Br
Quantity 25 g
Formula Weight 280.28
Percent Purity 99%
Physical Form Chunks and/or Powder
Chemical Name or Material Decyltrimethylammonium bromide, 99%
compliance-icons
Product Identifier
  • Decyltrimethylammonium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.