Learn More
Honeywell™ Deep Blue Winter Gloves
Click to view available options
Size:
7
8
9
10
11
Description
- Polyamide knitted glove
- Fully dipped blue foam nitrile coating
- Fully Lined with a polar fleece
- Self fastening system on the wrist
- The ergonomics seamless knit construction conforms better to the natural shape of the hand for greater strength and durability, superior confort
- The filament yarn reduce linting for the most sensitive applications and easily laundered for extended use
- The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The foam coating optimise the glove's breathability
- The nitrile foam improves the grip
- The knitted and self fasteing system gives the glove good support
- The nitrile coating on all the hand provides outstanding abrasion resistance and impermeability to oils
- The polar fleece brings a very good cold insulation
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Certifications/Compliance | EN 420-2003 Basic standards, EN 388:2016 Mechanical hazards, EN511-2006 Cold protection |
Description | 13 gauge |
Color | Blue |
Material | Nitrile |
Size | 11 |
Length (Metric) | 25 cm |