missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Deep Water Sampler

Operate this sampler with ease by simply lowering the sampler to the required depth and then pulling it out. Sampling Systems™ Deep Water Sampler is ideal for sampling at large depths.

Brand:  Sampling Systems™ 5079A-2500

Product Code. 15829361

  • £490.00 / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Supplied with 15m rope
  • Transparent plastic construction so the sample can be viewed inside
  • Ideal for fast stratified liquid sampling
  • Made from lightweight but robust plastic
  • Simple and easy to use in rivers, lakes or large deep containers
  • Takes a sample at the point you start pulling the handle up
  • As the sampler is lowered both the top and bottom valve opens allowing liquid to flow through
  • When you pull up the rope they valves close trapping the sample inside
Specifications

Specifications

Plastic
2.5 L
130 mm
15 meter rope
1.3 kg
340 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.