missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ DNA LoBind™ Plates
DNA LoBind Plates maximize sample recovery of nucleic acids by significantly reducing sample-to-surface binding. These plates are the ideal solution for sample preparation and long-term storage.
£239.00
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Polypropylene |
Sterility | Non-sterile |
Max. RCF | 6000 x g |
Purity or Quality Grade | PCR Clean (Non-sterile) |
Product Code | Brand | No. of Wells | Volume (Metric) Well | Color | Packaging | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | Volume (Metric) Well | Color | Packaging | Price | Quantity & Availability | ||
13427896
|
Eppendorf™
0030603303 |
96 | 350 μL | White, Clear | 80 Plates (5 Bags x 16 Plates) |
£239.00
Pack of 80 |
In Stock
Log in to see stock. |
||
Description
- Eppendorf LoBind material ensures optimized sample recovery for improved assay results.
- Free of surface coating (e.g. silicone) to minimize the risk of sample interference.
- Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
- High-contrast Unique OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors.
- RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
- Raised well rims and a smooth surface ensure reliable sealing.
Specifications
Autoclavable | |
Non-sterile | |
PCR Clean (Non-sterile) |
Polypropylene | |
6000 x g | |
Colorless |