Learn More
Eppendorf™ Deepwell Plate Rotor for A-2-DWP Benchtop Centrifuge
Use this rotor with Eppendorf™ model A-2-DWP Benchtop Centrifuge. Eppendorf™ Deepwell Plate Rotor for A-2-DWP Benchtop Centrifuge includes two deepwell plate buckets.
Brand: Eppendorf™ 5804740009
Code : 99
Additional Details : Weight : 3.70800kg
Description
- For use with: A-2-DWP Benchtop Centrifuge
Specifications
Deepwell Plate Rotor | |
4,000 rpm | |
2 buckets |
2,250 x g | |
A-2-DWP |