Learn More
Ohaus™ Defender™ 3000 Hybrid
OHAUS™ Defender™ 3000 Hybrid is ideal for general weighing and simple counting applications. Features a simple, yet rugged stainless steel design and 4X/IP66 stainless steel indicator with tactile keys, backlit LCD display, and built-in rechargeable battery.
Brand:  Ohaus™ 83997633
Product Code. 15374224
Description
Description
  • The IP66 stainless steel indicator is built to endure tough industrial elements.
  • The 58-hour rechargeable battery offers continuous operation for multi-day operations.
  • This tough industrial scale features a 304 stainless steel platform, stainless steel column and indicator bracket, painted tubular frame, and aluminum IP67 load cell.
  • Easy-to-read display and basic counting mode allow for simple operation for everyday basic industrial applications.
  • Offered with OIML/EC Type approvals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
58 Hours with Rechargeable Battery
IP65 stainless steel hinged front cover indicator, stainless steel column and wall bracket, 304 grade stainless steel platform with painted steel tubular frame, IP67 aluminum load cell
Backlit LCD
D32XW150BX-EU
-10°C to +40°C
740 x 500 x 920 mm
Easy access communication port with RS232 interface (included)
150.00 kg
20 g
IP65, NEMA 4X
Gram, Kilogram, Ounce, Pound, Pound:Ounce
28 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Ohaus™ Defender™ 3000 Hybrid > Model: D32XW150BX-EU