Learn More
OHAUS™ Defender™ 3000 - I-D33 Bench Scale
Bench Scales for Basic Industrial Applications
Brand: OHAUS™ 30684740
Additional Details : Weight : 10.37000kg Transport : UN number : 2800 Chem class : 8
Description
- Results are clearly shown using the large, bright backlit LCD display.
- In Checkweighing mode display changes color (yellow/green/red) to alert when within the target weight range.
- This industrial scale features a 304 stainless steel platform, powder-coated steel tubular frame, and aluminum load cell.
- Indicator features a stainless steel housing and mounting bracket.
- Features software for Parts Counting, Checkweighing and Totalization.
- Includes RS232 with optional USB Device or Ethernet for printing or data connectivity.
Specifications
100 Hours with Rechargeable Battery;360 Hours with Disposable Batteries | |
Standard RS232 and optional USB Device or Ethernet | |
60 kg | |
355 x 305 mm | |
D33P60B1R1 | |
Weighing, Checkweighing, Counting, Totalization. | |
10.9 kg |
6 C (LR14) Batteries (Not Included);AC Adapter (Included);Rechargeable Battery (Not Included) | |
ABS plastic indicator, 304-stainless steel platform, powder-coated steel tubular frame, aluminum load cell, non-slip adjustable rubber feet | |
7 Segment LCD with Multicolor Backlight | |
10 g | |
-10°C to +40°C, 85%RH, non-condensing | |
489 x 305 x 542 mm |