Learn More
OHAUS™ Defender™ 5000 Washdown - D52 Bench Scale, Models TD52XW QV
OHAUS™ Defender™ 5000 Series multifunctional bench scale is ideal for a multitude of applications. Built durably and equipped with advanced features such as maximum configurability print output, GMP/GLP data output, library/user management and multiple connectivity options.
£1790.00 - £2140.00
Specifications
Battery Life | 20 Hours with Rechargeable Battery |
---|---|
Connection Type | Easy access communication ports including RS232 and Micro SD slot (included), second RS232/RS485/USD device, Ethernet and WiFi/Bluetooth, Discrete I/O and Analog Output (sold separately as accessories); GLP/GMP data output with real-time clock |
Construction | 304-grade stainless steel platform and frame, stainless steel IP68 indicator, IP67 load cell, non-slip adjustable rubber feet |
Display Type | Backlit LCD, 256 x 80 pixels |
Protection Class | Stainless Seel IP68 Indicator, IP67 Load Cell |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
15908525
|
OHAUS™
30444827 |
D52XW300WQDV8-GB |
£1790.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15908585
|
OHAUS™
30461575 |
D52XW300WQDV8-M |
£1885.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15908655
|
OHAUS™
30471165 |
D52XW150WQV8-M |
£2140.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15958635
|
OHAUS™
30471150 |
D52XW60WQV8-GB |
£2045.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15968635
|
OHAUS™
30471151 |
D52XW150WQV8-GB |
£2045.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15988515
|
OHAUS™
30444825 |
D52XW60WQDV8-GB |
£1790.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15988575
|
OHAUS™
30461573 |
D52XW60WQDV8-M |
£1885.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15998515
|
OHAUS™
30444826 |
D52XW150WQDV8-GB |
£1790.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15998575
|
OHAUS™
30461574 |
D52XW150WQDV8-M |
£1885.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15998645
|
OHAUS™
30471164 |
D52XW60WQV8-M |
£2140.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- User-friendly model with an easy-to-use keypad, large backlit display and plain text on-screen prompts which guide you through operation.
- Durable construction with a stainless steel platform, frame, IP67 load cell and IP68 indicator.
- Defender meets NTEP/Measurement Canada or OIML/EC Type approvals (depending on region).
- Features common industrial modes and supports a 50,000-item library with 1,000 user profiles for easy data management.
- Incudes GMP/GLP with time/date for result traceability.
Specifications
20 Hours with Rechargeable Battery | |
304-grade stainless steel platform and frame, stainless steel IP68 indicator, IP67 load cell, non-slip adjustable rubber feet | |
Stainless Seel IP68 Indicator, IP67 Load Cell | |
773 x 610 x 1234 mm |
Easy access communication ports including RS232 and Micro SD slot (included), second RS232/RS485/USD device, Ethernet and WiFi/Bluetooth, Discrete I/O and Analog Output (sold separately as accessories); GLP/GMP data output with real-time clock | |
Backlit LCD, 256 x 80 pixels | |
-10°C to +40°C | |
36.800 kg |