missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Defined Trypsin Inhibitor
Defined Trypsin Inhibitor
Brand: Gibco™ R007100
Code : 50
Additional Details : Weight : 0.01050kg
Description
A sterile, phosphate-buffered saline solution (1X) containing purified soybean trypsin inhibitor. Animal origin-free.Specifications
ISO 13485 standards, FDA | |
Defined Trypsin Inhibitor Solution, 100mL, 1 X Concentration, Liquid, Sterile, Trypsin Inhibitor Reagent Solution, Purified Soybean Source, Used for Mammalian Cell Culture and Primary Cell Culture, –20°C Storage Temperature | |
1 X | |
Trypsin inhibitor | |
Mammalian cell culture, primary cell culture | |
For research use only |
–20°C | |
Mammalian Cells | |
Liquid | |
Sterile | |
100 mL | |
Purified soybean |
For Research Use Only. Not for use in diagnostic procedures.