Learn More
Deguelin, MedChemExpress
MedChemExpress Deguelin, a naturally occurring rotenoid, acts as a chemopreventive agent by blocking multiple pathways like PI3K-Akt, IKK-NF-κB, and MAPK-mTOR-survivin-mediated apoptosis. Deguelin binding to Hsp90 leads to a decreased expression of numerous oncogenic proteins, including MEK1/2, Akt, HIF1α, COX-2, and NF-κB.
Brand: Medchem Express HY-13425
Description
Deguelin, a naturally occurring rotenoid, acts as a chemopreventive agent by blocking multiple pathways like PI3K-Akt, IKK-NF-κB, and MAPK-mTOR-survivin-mediated apoptosis. Deguelin binding to Hsp90 leads to a decreased expression of numerous oncogenic proteins, including MEK1/2, Akt, HIF1α, COX-2, and NF-κB.Specifications
Deguelin | |
Light Yellow | |
C23H22O6 | |
(-)-Deguelin (-)-cis-Deguelin | |
O=C1[C@]2([H])[C@](COC3=CC(OC)=C(OC)C=C32)([H])OC4=C5C=CC(C)(C)OC5=CC=C14 | |
394.42 | |
Research | |
Cancer-Kinase/protease |
522-17-8 | |
Research | |
50 mg | |
DMSO : 100 mg/mL (253.54 mM; Need ultrasonic) | |
394.42 | |
98.0% | |
Solid |