Learn More
Thermo Scientific™ Deionizer Cartridges
Choose from an array of Thermo Scientific™ Deionizer Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
£84.90 - £273.00
Product Code | Brand | For Use With (Equipment) | Resin Type | Capacity | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Resin Type | Capacity | For Use With (Application) | Price | Quantity & Availability | |||||
12740666
|
Thermo Scientific™
D0760 |
Bantam™ Deionizer, B-Pure™ | Strong Anion Exchange | 1680 grains | Anion Removal |
£273.00
Each |
Estimated Shipment: 09-06-2025
Log in to see stock. |
|||||
14871651
|
Thermo Scientific™
D50273 |
Nanopure | Mixed Bed | Ultrapure |
£159.00
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||||
15342178
|
Thermo Scientific™
D8815 |
B-Pure™ | Activated Carbon | 5000 gal. (as CaCO3) | Organic Removal |
£119.00
1 unit |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
15352178
|
Thermo Scientific™
D8816 |
B-Pure™ | Mixed Bed | Ultrapure |
£140.00
1 unit |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
15362178
|
Thermo Scientific™
D8817 |
B-Pure™ | Oxygen Removal Resin | Oxygen Removal |
£189.00
1 unit |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||||
12331683
|
Thermo Scientific™
D8818 |
B-Pure™ | Ultrapure, Oxygen Removal | Ultrapure, Oxygen and Organic Removal |
£215.00
Each |
Estimated Shipment: 29-08-2025
Log in to see stock. |
||||||
15372178
|
Thermo Scientific™
D8825 |
B-Pure™ | Mixed Bed, Ultrapure, Oxygen and Organic Removal | Ultrapure and Oxygen Removal Resin |
£224.00
1 unit |
Estimated Shipment: 09-06-2025
Log in to see stock. |
||||||
11789428
|
Thermo Scientific™
D8831 |
Milli-Q Millipore Water Systems | Super-C Carbon | Organic Removal |
£84.90
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11799428
|
Thermo Scientific™
D8832 |
Milli-Q Millipore Water Systems | Mixed Bed | Ion Exchange, Ultrapure | N/A | |||||||
10071893
|
Thermo Scientific™
D0809 |
Bantam™ Deionizer, B-Pure™ | Mixed Bed | 875 grains |
£151.41
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
10589690
|
Thermo Scientific™
D0811 |
B-Pure™ | Porous Strong Ion Exchange | 30 g | Oxygen Removal |
£155.00
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
10041953
|
Thermo Scientific™
D0813 |
Bantam™ Deionizer, B-Pure™ | Activated Carbon | 5000 gal. | Organic Removal |
£122.00
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
12311278
|
Thermo Scientific™
D0815 |
Bantam™ Deionizer, B-Pure™ | Strong Cation Exchange | 3170 grains | Cation Removal |
£113.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11746189
|
Thermo Scientific™
D0832 |
Bantam™ Deionizer, B-Pure™ | Mixed Bed, Activated Carbon | 785 grains/1000 gal. | Ultrapure and Organic Removal |
£135.00
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
10451820
|
Thermo Scientific™
D0835 |
B-Pure™ | Macroreticular, Activated Carbon | 5000 gal. | Pretreatment |
£145.60
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
10297093
|
Thermo Scientific™
D0836 |
B-Pure™ | Macroreticular, Activated Carbon | 2000 gal. | Pretreatment Macropure |
£209.09
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
10074067
|
Thermo Scientific™
D50213 |
B-Pure™ | Mixed Bed | 550 grains | Ultrapure |
£133.00
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
|||||
15312168
|
Thermo Scientific™
D50214 |
B-Pure™ | Porous Strong Anion Exchange | 18 g | Oxygen Removal |
£147.00
1 unit |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
15322168
|
Thermo Scientific™
D50215 |
B-Pure™ | Activated Carbon | 5000 gal. | Organic Removal |
£111.00
1 unit |
Estimated Shipment: 09-06-2025
Log in to see stock. |
|||||
15297643
|
Thermo Scientific™
D50216 |
B-Pure™ | Oxygen Removal Mixed Bed | 275 grains/9 g | Oxygen and demineralization |
£140.00
Each |
Estimated Shipment: 08-08-2025
Log in to see stock. |
|||||
10125767
|
Thermo Scientific™
D50217 |
B-Pure™ | Mixed Bed, Activated Carbon | 275 grains/2000 gal. | Ultrapure and Organic Removal |
£140.00
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
11706289
|
Thermo Scientific™
D63112 |
B-Pure™ | Extruded Carbon | 5000 gal. | Organic Removal |
£149.00
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
12730746
|
Thermo Scientific™
D8809 |
B-Pure™ | Ultrapure, Oxygen Removal | 455 grains/30 g | Ultrapure and Organic Removal |
£151.00
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
10081893
|
Thermo Scientific™
D8811 |
B-Pure™ | Ultrapure, Oxygen and Organic Removal | 365 grains/2000 gal./12 g | Ultrapure, Oxygen and Organic Removal |
£146.00
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
10501673
|
Thermo Scientific™
D0803 |
Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as Indicated | Two Bed Ion Exchange | 1760 grains | Cartridge for high capacity; two-bed ion exchange; full size |
£149.76
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
Description
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
- Choice of full or half cartridges
Specifications
Deionizer Cartridge |