missing translation for 'onlineSavingsMsg'
Learn More

Scotch-Brite™ Delicate Surface Scouring Sponges

High quality, delicate surface cleaning sponges offer effective cleaning power on office furniture. Thanks to the softness of the sponge, it will not scratch. Very flexible, durable sponges that are easy to wash & rinse after use.

Brand:  Scotch-Brite™ 7100145508

194.75 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 17138760

  • £225.00 / Pack of 100
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High quality delicate surface cleaning sponges
  • Softness of the sponge means it won't scratch surfaces
  • Handy size for a good grip and excellent durability
  • Easy to rinse/wash after use
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sponges
133 x 70 mm
Office Furniture
10 Pk.
Beige, Red
Cleaning Delicate Surfaces
133 mm
70 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.