Learn More
Cole-Parmer™ Delrin™ Manual Pinch Valve
Brand:  Cole-Parmer™ 98002-10
 View more versions of this product
Product Code. 11747288
  • £65.60 / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Manual Pinch Valve
Fits 5/16 in. to 3/8 in. Tube O.D.
1 Pack
Delrin
Ideal for use in pharmaceutical and food industries where eliminating contamination is critical