Learn More
Fisherbrand™ Delta™ Cell Spreaders
Spread cultures evenly and carefully. Delta™ Cell Spreaders have a flexible handle with a domed spreading bar to help reduce agar tears.
Brand: Fisherbrand™
100.32 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 41
Additional Details : Weight : 2.34900kg
Description
- Flexible rippled handle for better control.
- Domed spreading bar headpiece helps reduce agar tears.
- Gamma sterilized and individually wrapped.
- 500 disposable spreaders per pack.
Specifications
Single-use | |
Sterile | |
Spreader | |
172 mm |
6.8 in. | |
Delta | |
ABS |