missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Deltasafe Flame-Retardant M1 Polypropylene Coveralls with Hood
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Comfortable with Breathability Equal to a T-Shirt
- Flame-Retardant and Antistatic Certified (EN 533 and EN 1149-1)
- M1 and Radioactive Particle Certified (NF P92-507 and EN 1073-2)
- Protection Level: General Requirements, Type 5 and Type 6 (EN 340, EN 13982-1, and EN 13034), EC Category PPE: 3
- Recommended Industries: Chemical, Oil and Gas, Nuclear
- Hazards: Airborne Particulate Contamination, Chemical Splash
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Size | 2X-Large |
Gender | Unisex |
Material | Polypropylene |
Color | White |
Disposable | Single-use |
Ankle Style | Elastic |
Cuff Style | Elastic |
Hood Type | Elastic |
Sterility | Non-sterile |
Protection Class | Protection Type: 5, 6 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'