missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Denaturing Lysis Buffer
£31.96 - £34.96
Specifications
Format | Liquid |
---|---|
Product Type | Cell Lysis Reagent |
pH | 7.4 |
Content And Storage | Ambient temperatures |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
15442959
|
Thermo Scientific™
J60132.AK |
250mL |
£31.96
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15452959
|
Thermo Scientific™
J60132.AP |
500mL | N/A | N/A | N/A | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Liquid | |
Cell Lysis Reagent | |
7.4 | |
Ambient temperatures |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Denaturing Lysis Buffer