missing translation for 'onlineSavingsMsg'
Learn More

PEPperCHIP™ Dengue Virus Type 3 Proteome Microarray

Dengue Virus Type 3 Proteome Microarray with 3390 virus specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.

Brand:  PEPperCHIP™ SPC.006.001

Additional Details : Weight : 0.02200kg

Product Code. 16397748

  • £482.30 / Each
Estimated Shipment: 22-10-2024
to see stock.
Add to basket

Description

Description

  • Microarray Content: Genome polyprotein (strain Philippines/H87/1956, UniProt ID P27915) translated into overlapping peptides
  • Peptide Length/Overlap: 15 aa/ 14 aa
  • Number of Peptides: 3390 printed in duplicate and HA, polio and c-Myc control peptides
Specifications

Specifications

PEPperSlide™ PEGMA/PMMA
Antibody validation, epitope mapping, serum screening, biomarker discovery
Microscopic Glass
Individual barcode for each microarray
1 Slide
1 mm
Transparent
75.4 x 25 mm
High-density Peptide Microarray
Non-sterile
1 array copy per microarray
Combinatorial peptide synthesis by amino acid laser printing
ISO 9001:2015
RUO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers