Learn More
Denhardt' Reagent (50X Solution/Molecular Biology), Fisher BioReagents
Brand: Fisher Bioreagents BP515-5
Code : A0
Additional Details : CAS Number : 7732-18-5 Weight : 0.06000kg
Specifications
0.60 to 0.80 AU at 278nm | |
Colorless | |
26873-85-8,9003-39-8,9048-46-8 | |
0°C | |
<1% each of Ficoll™, Polyvinylpyrrolidone, and Bovine Albumin; and Water. Each 5mL of 50X Solution contains 50mg each of Ficoll™, Polyvinylpyrrolidone, and Bovine Albumin. | |
Report (at 25°C) | |
5 mL |
Denhardt's Reagent | |
50X Solution | |
100°C | |
Molecular Biology | |
Amber Glass | |
5 to 7 | |
Liquid |