missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Dense Storage Options
Double your cryobank capacity and cut storage costs by 50%. Our dense storage options fit standard cryogenic storage boxes that work with the freezer racks you currently own.
£80.56 - £1085.00
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10564985
|
Thermo Scientific™
374001 |
Cryobox divider, 13 x 13 grid |
£163.00
Pack of 24 |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||||
10233091
|
Thermo Scientific™
5050-0001 |
Polycarbonate CryoBox, Undivided |
£140.00
Pack of 4 |
In Stock
Log in to see stock. |
|||||
15323584
|
Thermo Scientific™
5050-0169 |
Polycarbonate CryoBox with 13 x 13 divider |
£320.00
Pack of 24 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||||
15201866
|
Thermo Scientific™
374115 |
1.0mL Nunc Cryobank Tubes, Blank, with Caps, Sterile, Loose Packed |
£1085.00
Pack of 845 |
In Stock
Log in to see stock. |
|||||
11787064
|
Thermo Scientific™
369640 |
Cardboard Cryo box with 13 x 13 divider |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15183113
|
Thermo Scientific™
369678 |
Cryobank Universal Picker/Pen |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Double your cryobank storage capacity and cut storage costs by 50%. The Thermo Scientific™ Dense Storage Options use standard CryoBoxes™ with 13 x 13 tube separators and fit 169 1mL tubes in a single CryoBox™. These standard-sized boxes are compatible with existing standard-sized lab freezer racks.
- Choose a standard-sized cardboard or polycarbonate box
- Insert the 13x13 box divider
- Load the box with 169, 1mL Cryobank storage tubes
- Slide the box into your existing standard-sized CryoBox racks
- Suitable for mechanical freezer or vapor phase liquid nitrogen storage