Learn More
Density Standard, ASTM D 4052; ISO 12185 (Nominal Value: 0.73576 g/mL, 15 °C), Paragon Scientific™
Density Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Density according to ASTM D4052. Alternative test methods are IP 365 and ISO 12185.
Brand: Paragon Scientific CRM-DEGA
Additional Details : Weight : 0.25000kg Transport : UN number : 1203 Chem class : 3 Pack group : II
Description
- Certified Reference Material for Density
- Manufactured in accordance with ASTM / EN or IP test methods
- Certified via Round Robin
- Supplied with MSDS.
- Tamper evident security packaging.
- 12 month shelf life.
- Manufactured in the United Kingdom
Specifications
UN 1203 |
Safety and Handling
- Certified Reference Material Density Standard, Gasoline
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Carcinogenicity Category 1B
- Flammable liquid Category 1
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H224-Extremely flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H361-Suspected of damaging fertility or the unborn child.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
- P391-Collect spillage.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : gasoline
Nominal values quoted. Certified Values will vary from batch to batch.