missing translation for 'onlineSavingsMsg'
Learn More

Density Standard, ASTM D 4052; ISO 12185 (Nominal Value: 0.83294g/mL, 15 °C), Paragon Scientific™

Density Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Density according to ASTM D4052. Alternative test methods are IP 365 and ISO 12185.

Brand:  Paragon Scientific CRM-DEGO


Product Code. 16060182

  • £222.00 / 250mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Certified Reference Material for Density
  • Manufactured in accordance with ASTM / EN or IP test methods
  • Certified via Round Robin
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • 12 month shelf life.
  • Manufactured in the United Kingdom
Specifications

Specifications

UN 1202
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Density Standard

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : diesel

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Nominal values quoted. Certified Values will vary from batch to batch.