missing translation for 'onlineSavingsMsg'
Learn More

Density Standards (100 °C), Paragon Scientific™

Paragon ISO 17025 / ISO 17034 certified Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 15°C to 150°C

Brand:  Paragon Scientific DEN100-01


 View more versions of this product

Product Code. 13420539

  • £70.40 / 60mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in accordance with primary level ASTM D1480 methodology
  • Ensures full compliance to ASTM and IP test method protocols.
  • Supplied in tamper evident packaging with a 2 year shelf life.
  • Fully traceable to international standards.
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • Manufactured in the United Kingdom
Specifications

Specifications

60 mL
0.768 g/mL (100°C)
Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • DEN100-01

Signal Word
  • Danger

Hazard Category
  • Aspiration toxicity Category 1

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.

Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

Supplemental information
  • MIXTURE LIST-Contains : Kerosine sweetened

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Nominal values quoted